Learn More
Mettler Toledo™ Automatic Feeder LV12
Catalog No. 01919120 Shop All METTLER TOLEDO Products
Change view
Click to view available options
Automatic Feeder
Catalog No.
01-919-120 Automatic Feeder
1 options
Catalog No. 01-919-120 Supplier Mettler Toledo™ Supplier No. 30374389
Add to Cart
Add to Cart
Request a Quote
Feeds tablets and other small, unattended parts piece by piece into the weighing pan
  • Automatic Feeder LV12
  • Reliable Item Separation
  • Automate Weighing Process for Tablets
  • Small, unattended parts are reliably fed piece by piece into the weighing pan
  • Speed up statistical quality control
  • Shortens the time required for weighing large samples of tablets
  • Process is controlled by connected SQC balance or device
  • Controlled interface to the balance ensures synchronized feeding of items
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) XPR Balance
For Use With (Application) Automatic Weighing
Type Automatic Feeder
Autoclavable Not Autoclavable