Learn More
Electron Microscopy Sciences Automatic Pipette Washing Set
SDP
Catalog No. 501931097
Encompass_Preferred
Solve the problem of washing pipettes quickly, safely and with little effort. To use allow dirty pipettes to soak in the pipette basket in the pipette jar and when ready the basket is transferred to the washer where a constant flow of tap water rinses the pipettes clean.
Catalog No. 50-193-1097 Supplier Electron Microscopy Sciences Supplier No. 6437731
Solve the problem of washing pipettes quickly, safely and with little effort. To use allow dirty pipettes to soak in the pipette basket in the pipette jar and when ready the basket is transferred to the washer where a constant flow of tap water rinses the pipettes clean.