Learn More
Anton Paar Automatic viscometer with Simple Fill funnel—pour and go, no pipette or syringe needed. 150% higher throughput vs. glass capillary viscometers. Results in ASTM D7042 and D445 (bias-corrected) with in-place cleaning and drying.
Catalog No. 5000580466
Encompass_Preferred
Test Methods: ASTM D7042, D445 bias corrected, ISO 23581, EN 16896Measuring Range:Viscosity: 0.3 mm²/s to 5,000 mm²/sTemperature: +15 °C to +100 °C (one temperature, second temperature optional)Viscosity repeatability** and reproducibility**0.1 % and 0.35 %Temperature repeatability: 0.005 °CTemperature reproducibility: 0.03 °C from +15 °C to +100 °CSample volume min. / typical: 3.5 mL / 8 mLSolvent volume min. / typical: 5 mL / 10 mLMax. sample throughput: 21 samples per hourOptional upgrades: Second temperatureIntegrate Simple Fill Filling Device
Catalog No. 50-005-80466 Supplier Anton Paar Supplier No. 105011
Please contact your sales representative for a quote or further ordering instructions.
Test Methods: ASTM D7042, D445 bias corrected, ISO 23581, EN 16896Measuring Range:Viscosity: 0.3 mm²/s to 5,000 mm²/sTemperature: +15 °C to +100 °C (one temperature, second temperature optional)Viscosity repeatability** and reproducibility**0.1 % and 0.35 %Temperature repeatability: 0.005 °CTemperature reproducibility: 0.03 °C from +15 °C to +100 °CSample volume min. / typical: 3.5 mL / 8 mLSolvent volume min. / typical: 5 mL / 10 mLMax. sample throughput: 21 samples per hourOptional upgrades: Second temperatureIntegrate Simple Fill Filling Device