Learn More
Chemglass Life Sciences Automation conductive pipette tips, 1000 μL - for Hamilton automation systems, pre-sterilized, 96/Pack
SDP
Catalog No. 502733249
Encompass_Preferred
Chemglass Automation conductive filter tips are high-volume conductive pipette tips engineered for automated liquid handling on Hamilton workstations. They provide uniform conductivity, aerosol protection, and are supplied pre-sterilized in peelable Tyvek and blister packaging to maintain sterility and traceability.
  • 1000 μL nominal volume.
  • Medical grade black conductive polypropylene construction.
  • Pre-sterilized by electron beam.
  • 96 tips per pack (rack), 2,304 tips per case.
  • Packed in peelable Tyvek films and blister packs for sterility.
  • Manufactured in a 100,000-grade clean plant and tested for DNA, RNA, proteases, and pyrogens.
  • Designed for high-throughput automated pipetting with uniform conductivity and barcode traceability.
Catalog No. 50-273-3249 Supplier Chemglass Life Sciences Supplier No. CLS3499A1002
Chemglass Automation conductive filter tips are high-volume conductive pipette tips engineered for automated liquid handling on Hamilton workstations. They provide uniform conductivity, aerosol protection, and are supplied pre-sterilized in peelable Tyvek and blister packaging to maintain sterility and traceability.
  • 1000 μL nominal volume.
  • Medical grade black conductive polypropylene construction.
  • Pre-sterilized by electron beam.
  • 96 tips per pack (rack), 2,304 tips per case.
  • Packed in peelable Tyvek films and blister packs for sterility.
  • Manufactured in a 100,000-grade clean plant and tested for DNA, RNA, proteases, and pyrogens.
  • Designed for high-throughput automated pipetting with uniform conductivity and barcode traceability.