Learn More
Chemglass Life Sciences Chemglass Automation Conductive Filter Tips, 200 μl, for Tecan, Pre-sterilized, 96/pack
SDP
Catalog No. 502733241
Encompass_Preferred
Chemglass Automation conductive filter tips are 200 μl, pre-sterilized pipette tips designed for high-throughput automated liquid-handling workflows. Made from medical-grade conductive polypropylene and produced in a controlled cleanroom environment, these tips include internal filters and are packaged to minimize contamination during storage and use.
  • Made from medical-grade black conductive polypropylene.
  • Pre-sterilized by electron beam to remove biological contaminants.
  • Manufactured in a 100,000-grade clean plant.
  • Free of DNA, RNA, protease, and pyrogen.
  • Designed for high-throughput automated liquid-handling systems.
  • Packaged 96 tips per pack and 2,304 tips per case.
  • Wrapped in peelable Tyvek films and blister packs for contamination control.
  • Intended for 200 μl volume with internal filter protection.
Catalog No. 50-273-3241 Supplier Chemglass Life Sciences Supplier No. CLS3499A200
Add to Cart
Add to Cart
Chemglass Automation conductive filter tips are 200 μl, pre-sterilized pipette tips designed for high-throughput automated liquid-handling workflows. Made from medical-grade conductive polypropylene and produced in a controlled cleanroom environment, these tips include internal filters and are packaged to minimize contamination during storage and use.
  • Made from medical-grade black conductive polypropylene.
  • Pre-sterilized by electron beam to remove biological contaminants.
  • Manufactured in a 100,000-grade clean plant.
  • Free of DNA, RNA, protease, and pyrogen.
  • Designed for high-throughput automated liquid-handling systems.
  • Packaged 96 tips per pack and 2,304 tips per case.
  • Wrapped in peelable Tyvek films and blister packs for contamination control.
  • Intended for 200 μl volume with internal filter protection.