Learn More
Chemglass Life Sciences Automation Conductive Filter Tips, 300 μl - For Hamilton Microlab, STAR, and NIMBUS Systems; Pre-sterilized, Black; 96/Pack
SDP
Catalog No. 502733239
Encompass_Preferred
Conductive 300 μl automation pipette tips designed for Hamilton automated liquid handlers. These pre-sterilized filter tips are molded from medical-grade conductive polypropylene to reduce static and protect samples during automated dispensing. Packaging supports nest and blister formats for high-throughput workflows.
  • 300 μl nominal volume.
  • Conductive medical-grade polypropylene, black.
  • Pre-sterilized by electron beam.
  • Filter design reduces aerosol contamination.
  • Packed 96 per pack, 2,304 per case.
  • Compatible with nest and blister automation handling.
  • Manufactured under cleanroom conditions and quality tested for nucleic acid and protease contamination.
Catalog No. 50-273-3239 Supplier Chemglass Life Sciences Supplier No. CLS3499A302
Conductive 300 μl automation pipette tips designed for Hamilton automated liquid handlers. These pre-sterilized filter tips are molded from medical-grade conductive polypropylene to reduce static and protect samples during automated dispensing. Packaging supports nest and blister formats for high-throughput workflows.
  • 300 μl nominal volume.
  • Conductive medical-grade polypropylene, black.
  • Pre-sterilized by electron beam.
  • Filter design reduces aerosol contamination.
  • Packed 96 per pack, 2,304 per case.
  • Compatible with nest and blister automation handling.
  • Manufactured under cleanroom conditions and quality tested for nucleic acid and protease contamination.