Learn More
Chemglass Life Sciences Chemglass Automation Conductive Pipette Tips, 50 μL, Tecan-compatible, Pre-sterile
SDP
Catalog No. 502733264
Encompass_Preferred
Chemglass 50 μL conductive automation pipette tips are designed for high-throughput liquid-handling on Tecan systems. Pre-sterilized and packaged in peelable Tyvek films and blisters, these tips provide uniform conductivity, low contamination risk, and consistent performance for genomics, proteomics, and biopharmaceutical workflows.
  • Volume: 50 μL.
  • Compatibility: Tecan liquid-handling systems.
  • Packaging: 96 tips per pack; 2,304 tips per case; wrapped in peelable Tyvek films and blisters.
  • Sterilization: pre-sterilized by electron beam.
  • Material: medical-grade black conductive polypropylene.
  • Manufacturing: produced in a 100,000-grade clean plant.
  • Quality control: batch-tested for uniform conductivity, resistance, and CV values.
  • Contamination control: tested free of DNA, RNA, proteases, and pyrogens.
Catalog No. 50-273-3264 Supplier Chemglass Life Sciences Supplier No. CLS3499A050
Add to Cart
Add to Cart
Chemglass 50 μL conductive automation pipette tips are designed for high-throughput liquid-handling on Tecan systems. Pre-sterilized and packaged in peelable Tyvek films and blisters, these tips provide uniform conductivity, low contamination risk, and consistent performance for genomics, proteomics, and biopharmaceutical workflows.
  • Volume: 50 μL.
  • Compatibility: Tecan liquid-handling systems.
  • Packaging: 96 tips per pack; 2,304 tips per case; wrapped in peelable Tyvek films and blisters.
  • Sterilization: pre-sterilized by electron beam.
  • Material: medical-grade black conductive polypropylene.
  • Manufacturing: produced in a 100,000-grade clean plant.
  • Quality control: batch-tested for uniform conductivity, resistance, and CV values.
  • Contamination control: tested free of DNA, RNA, proteases, and pyrogens.