Learn More
Thermo Scientific™ Automation Reservoirs, with removable splash baffles
GREENER_CHOICE
Catalog No. 14387036 Shop All Thermo Scientific Products
Change view
Click to view available options
Channels:
96
384
Sterility:
Non-sterile
Sterile
Description:
Deepwell
Material:
Plastic
Polypropylene
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Channels Sterility Description Material
14-387-036 384 Sterile   Polypropylene
14-387-032 96 Sterile   Polypropylene
14-387-034 96 Sterile Deepwell Polypropylene
14-387-035 384 Non-sterile   Polypropylene
14-387-031 96 Non-sterile   Polypropylene
14-387-033 96 Non-sterile Deepwell Plastic
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 14-387-036 Supplier Thermo Scientific™ Supplier No. 1064157
  • $588.50
    $379.65 / Case of 20
    Save $208.85
    35% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
Handle reagents on automated systems simply and economically.
Thermo Scientific™ Automation Reservoirs provide standard SBS dimensions ensuring compatibility with plate stackers and handlers, which enables you to easily handle reagents on automated systems economically. Reservoirs are disposable with no need for washing, allowing more reservoirs to be used per run. Built-in grooves in reservoir bottom reduce reagent waste and allow consistent aspiration.
Features:
  • Standard microplate dimensions enables reservoirs to be used in plate handlers or stackers
  • Hydrophilic surface so liquid spreads out evenly with minimal reagent needed to cover bottom
  • Polypropylene construction offers excellent chemical resistance
  • Available individually wrapped for sterile use
Removable Splash Baffles:
  • Prevent reagents from splashing out when being moved on robotic decks
  • Enable reservoirs to be moved faster so they can also be filled closer to the top with reagent
  • Can be removed to increase reservoirs capacity
TRUSTED_SUSTAINABILITY
Specifications
Description  
Material Polypropylene
Sterility Sterile
Type Reservoir
Includes Removable splash baffles
Packaging Individually Wrapped
Channels 384