Learn More
Anton Paar Automation Solution for Nanoparticle Analysis
Catalog No. 5000580512
Encompass_Preferred
Unattended sequences of up to 80 cuvettesMulti-cuvette studies incl. molecular mass workflowsSeamless control in Kalliope: setup, scheduling, and evaluationCollaborative robot: no safety enclosure neededCompatibility with disposable, glass, quartz, and Omega cuvettes.
Catalog No. 50-005-80512 Supplier Anton Paar Supplier No. 296391
Please contact your sales representative for a quote or further ordering instructions.
Unattended sequences of up to 80 cuvettesMulti-cuvette studies incl. molecular mass workflowsSeamless control in Kalliope: setup, scheduling, and evaluationCollaborative robot: no safety enclosure neededCompatibility with disposable, glass, quartz, and Omega cuvettes.