Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Axygen™ Automation Tips for Agilent™/Velocity11 and Bravo, 70 μL
GREENER_CHOICE

Catalog No. 14222895 Shop All Axygen Products
Change view
Click to view available options
Sterility:
Non-sterile
Sterile
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sterility
14-222-895 Non-sterile
14-222-896 Sterile
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 14-222-895 Supplier Axygen™ Supplier No. VT38470ULR
Add to Cart
Add to Cart
Request a Quote

Compatible with automated liquid handling systems. Axygen™ Automation Tips for Agilent™/Velocity11 and Bravo, 70μL are non-filtered pipette tips in 384-well racks. Constructed of clear polypropyle under strict controls to ensure quality. Options include sterile, non-sterile and double labeled tips.

  • Manufactured to stringent specifications under strict process controls
  • Assembled via automation to ensure consistent performance and quality
  • Certified RNase-/DNase-free and nonpyrogenic
  • 384-well racks of non-filtered pipette 70μL tips
  • Compatible with Agilent™/Velocity VPrep™ and Bravo 384ST, 96ST and 16ST head
  • Case includes 50 racks (19,200 tips).
TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 70 μL
Filtered Non-filtered
Sterility Non-sterile
Color Clear
Clean Claims DNase, RNase Free
For Use With (Application) Non-pyrogenic
Format Rack
Tip Style Automation, Round
Packaging SBS Rack
Quantity 50 Racks/Cs.
For Use With (Equipment) Agilent/Velocity VPrep 16ST, Agilent/Velocity VPrep 96ST, Agilent/Velocity VPrep 384ST, Agilent Bravo 384ST
Material Polypropylene
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.