Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Automation Tips for Beckman™ Liquid Handling Systems with AP384 Dispense Head
GREENER_CHOICE

Catalog No. 21935261 Shop All Thermo Scientific Products
Change view
Click to view available options
Sterility:
Sterile
Filtered:
Filtered
Non-filtered
Fit for Purpose:
Sterile
Sterile and PCR Ready
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sterility Filtered Fit for Purpose
21-935-261 Sterile Non-filtered Sterile
13-811-180 Sterile Filtered Sterile and PCR Ready
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 21-935-261 Supplier Thermo Scientific™ Supplier No. 935261
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

Increase pipetting performance with Thermo Scientific™ Automation Tips for Beckman™ Liquid Handling Systems, which undergo a rigorous 15-point quality control program to ensure consistency.

Increase pipetting performance with Thermo Scientific™ Sterile Pipette Tips for Beckman™ Liquid Handling Systems, which undergo a rigorous 15-point quality control program to ensure consistency. These automation tips provide accurate and precise performance due to superior tip straightness and a low coefficient of variation. The pipette tips are validated on the actual workstations for which they are created.

Features:
  • Available with Thermo Scientific™ ART™ filter technology, low retention and extended length
  • Lot-tested during production on actual workstation for which they are created
  • Extended length tips compatible with 96-, 384-, and 1536-deep well pates
  • Low percentage coefficient of variation (%CV)
TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 30 μL
Filtered Non-filtered
Sterility Sterile
Color Clear
Tip Style Extended Length
For Use With (Equipment) Beckman Coulter Liquid Handling Systems
Product Line Beckman
Fit for Purpose Sterile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.