Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Automation Tips for Beckman™ Liquid Handling Systems with MP20 and MP200 Dispense Heads
GREENER_CHOICE

Catalog No. 21402325
Change view
Click to view available options
Volume (Metric):
20 μL
130 μL
200 μL
Filtered:
Filtered
Non-filtered
Sterility:
Non-sterile
Sterile
Fit for Purpose:
Certified Quality
Sterile
Sterile and PCR Ready
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Filtered Sterility Fit for Purpose
21-402-325 20 μL Non-filtered Sterile Sterile
21-402-327 130 μL Filtered Sterile Sterile and PCR Ready
21-402-328 200 μL Non-filtered Sterile Sterile
21-402-329 200 μL Non-filtered Non-sterile Certified Quality
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 21-402-325 Supplier Thermo Scientific™ Supplier No. 915261
Add to Cart
Add to Cart
This item is not returnable. View return policy

Increase pipetting performance with Thermo Scientific™ Automation Tips for Beckman™ Liquid Handling Systems, which undergo a rigorous 15-point quality control program to ensure consistency.

Increase pipetting performance with Thermo Scientific™ Sterile Pipette Tips for Beckman™ Liquid Handling Systems, which undergo a rigorous 15-point quality control program to ensure consistency. These automation tips provide accurate and precise performance due to superior tip straightness and a low coefficient of variation. The pipette tips are validated on the actual workstations for which they are created.

Features:
  • Available with Thermo Scientific™ ART™ filter technology, low retention and extended length
  • Lot-tested during production on actual workstation for which they are created
  • Extended length tips compatible with 96-, 384-, and 1536-deep well plates
  • Low percentage coefficient of variation (%CV)
TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 20 μL
Filtered Non-filtered
Sterility Sterile
Color Clear
Format Rack
Tip Style Extended Length
For Use With (Equipment) Beckman Coulter Liquid Handling Systems
Product Line Beckman
Fit for Purpose Sterile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.