Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Automation Tips for Caliper™ Zymark™ Liquid Handling Workstations
GREENER_CHOICE

Catalog No. 21402304
Change view
Click to view available options
Volume (Metric):
50 μL
80 μL
100 μL
150 μL
200 μL
Filtered:
Filtered
Non-filtered
Sterility:
Non-sterile
Sterile
Fit for Purpose:
Certified Quality
Sterile
Sterile and PCR Ready
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Filtered Sterility Fit for Purpose
21-402-304 100 μL Non-filtered Sterile Sterile
10-371-033 50 μL Filtered Sterile Sterile and PCR Ready
10-371-032 50 μL Non-filtered Sterile Sterile
10-371-030 50 μL Non-filtered Non-sterile Certified Quality
10-371-031 80 μL Filtered Sterile Sterile and PCR Ready
10-371-035 100 μL Non-filtered Non-sterile Certified Quality
21-402-300 150 μL Filtered Sterile Sterile and PCR Ready
10-371-034 200 μL Non-filtered Sterile Sterile
21-402-302 200 μL Non-filtered Non-sterile Certified Quality
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. 21-402-304 Supplier Thermo Scientific™ Supplier No. 921261
Only null left
Add to Cart
Add to Cart

Increase pipetting performance with these automation tips, which undergo a rigorous 15-point quality control program to ensure accuracy and consistency.

Increase pipetting performance with Thermo Scientific™ Automation Tips for Caliper™ Zymark™ Liquid Handling Workstations, which undergo a rigorous 15-point quality control program to ensure consistency. Their superior tip straightness and a low coefficient of variation provide accurate and precise performance. Compatible with: Caliper/Zymark RapidPlate, Caliper Presto, Allegro, Zephyr and SciClone.

Features:

TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 100 μL
Filtered Non-filtered
Sterility Sterile
Color Clear
Format Rack
Tip Style Automation
For Use With (Equipment) Zymark/Caliper Liquid Handling Workstations
Product Line Caliper Zymark
Fit for Purpose Sterile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.