Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Automation Tips for Molecular Devices™ Pipetting System

Catalog No. p-4525570
Change view
Click to view available options
Volume (Metric):
20 μL
Sterility:
Sterile
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Sterility
14-754-337 20 μL Sterile
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14-754-337 Supplier Thermo Scientific™ Supplier No. 937261

Increase pipetting performance through superior tip straightness and low %CV with these pipette tips for the Molecular Devices system.

Increase pipetting performance with Thermo Scientific™ Automation Tips for Molecular Devices™ Pipetting System, which undergo a rigorous 15-point quality control program to ensure consistency. Their superior tip straightness and a low coefficient of variation provide accurate and precise performance. Compatible with: Molecular Device FLIPR with Type A CCS 384-well head and Type B Liberty 384-well head.

  • Available in non-sterile, sterile and with the Thermo Scientific™ ART™ filter technology
  • Low percentage coefficient of variation (%CV)
TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 20 μL
Filtered Non-filtered
Sterility Sterile
Color Clear
Format Rack
Tip Style Automation
For Use With (Equipment) Molecular Devices FLIPR Type A CCS 384-well Head
Product Line Molecular Devices

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.