Learn More
Axygen™ Automation Tips for PerkinElmer™, 96-Well
GREENER_CHOICE
Catalog No. 14222371 Shop All Axygen Products
Change view
Click to view available options
Volume (Metric):
20 μL
50 μL
200 μL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
14-222-371 50 μL
14-222-365 20 μL
14-222-361 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 14-222-371 Supplier Axygen™ Supplier No. PK50R
Get Up to $2,500 in Products with Purchase
View all promotions
Add to Cart
Add to Cart
Ideal for Janus™ MiniTrak, PlateTrak™ and Evolution P3. Axygen™ Automation Tips for PerkinElmer™, 96-Well are constructed of clear polypropylene. Available in sterile/non-sterile, filtered/non-filtered options, from 20 to 200 μL.
Assembled via automation under strict process controls to ensure consistent performance and quality
  • Robotic tips use no mold-releasing agents
  • Nucleic acid, nuclease and endotoxin-safe for complete protection against contamination
  • Tips are certified RNase-/DNase-free and nonpyrogenic
  • Compatible with PerkinElmer™ MDT-96 P235, MDT-96 P50 and MDT-384 30μL head for:
    • Janus™
    • Evolution P3
    • Minitrak
    • PlateTrak™
Multiple options
  • Presterilized and non-sterile tips
  • With or without aerosol-resistant polyethylene filters
  • Wide bore series for working with viscous materials, genomic DNA or delicate cell cultures
  • Available in a range of volumes: 20, 30, 50, 180 and 200μL
Order Info
Rack: 96 Tips/Rack, 10 Racks/Pack, 5 Packs/Case
Compliance
Axygen products are manufactured in a clean, controlled state of the art facility and certified RNase-, DNase- and endotoxin-safe
TRUSTED_SUSTAINABILITY
Specifications
Volume (Metric) 50 μL
Filtered Non-filtered
Sterility Non-sterile
Color Clear
Rack Color Green
Certifications/Compliance ANSI
Clean Claims DNase, Rnase Free
For Use With (Application) Non-pyrogenic, used for stringent specifications under strict process controls, nucleic acid, nuclease and endotoxin-safe for complete protection against contamination
Format Rack
Tip Style Automation, Round
Packaging SBS Rack
Quantity 50 Racks/Cs.
For Use With (Equipment) PerkinElmer Janus, PerkinElmer Evolution P3, PerkinElmer MiniTrak, PerkinElmer PlateTrak, PerkinElmer MDT-96 P235, PerkinElmer MDT-96 P50, PerkinElmer MDT-384 30μL
Material Polypropylene
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov