Learn More
Thermo Scientific™ Automation Tips for Tecan™ Liquid Handling Platforms with Te-MO 96 Multichannel Arm
Shop All Thermo Scientific Products

Description
Increase pipetting performance with Thermo Scientific™ Automation Tips for Tecan™ Liquid Handling Platforms, which undergo a rigorous 15-point quality control program to ensure consistency. Their superior tip straightness and a low coefficient of variation provide accurate and precise performance. Compatible with: Tecan Te-MO, Aquarius and Freedom EVO with Te-MO 96 head.
- Available as standard, filtered and/or liquid sensing
- Certified free of RNase, DNase, DNA, endotoxin, bioburden and pyrogen
- Lot-tested during production on actual workstation for which they are created
- Fully conductive, providing increased pipetting performance
- Low percentage coefficient of variation (%CV)
Specifications
Specifications
| Volume (Metric) | 150 μL |
| Filtered | Filtered |
| Sterility | Sterile |
| Color | Clear |
| Format | Rack |
| Tip Style | Automation |
| For Use With (Equipment) | Tecan Freedom EVO, Tecan TeMo, Tecan Aquarius |
| Product Line | Tecan |
| Fit for Purpose | Sterile and PCR Ready |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.