Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Axygen™ Automation Tips for Tecan™, Zymark™, Biotek™ and Dynamic Devices

Catalog No. 14222069 Shop All Axygen Products
Change view
Click to view available options
Sterility:
Non-sterile
Sterile
Volume (Metric):
50 μL
100 μL
180 μL
200 μL
205 μL
Filtered:
Filtered
Non-filtered
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sterility Volume (Metric) Filtered
14-222-069 Non-sterile 200 μL Non-filtered
14-222-074 Non-sterile 50 μL Non-filtered
14-222-077 Sterile 180 μL Filtered
14-222-075 Sterile 50 μL Non-filtered
14-222-079 Sterile 205 μL Filtered
14-222-080 Sterile 50 μL Filtered
14-222-076 Sterile 100 μL Filtered
14-222-071 Sterile 200 μL Non-filtered
14-222-078 Non-sterile 205 μL Filtered
14-222-066 Non-sterile 100 μL Non-filtered
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Catalog No. 14-222-069 Supplier Axygen™ Supplier No. EV200R
  • PRODUCT PROMOTION
    $219.50
    $135.00 / Pack of 960

    Save $84.50

    38% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
  • PRODUCT PROMOTION
    $989.50
    $675.00 / Case of 5 PK

    Save $314.50

    32% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ensure consistent performance and quality. Axygen™ Automation Tips for Tecan™, Zymark™, Biotek™ and Dynamic Devices are constructed of clear polypropylene in 96-well racks. Available in nonsterile/sterile, non-filtered/filtered, wide-bore and bottom-labeled options with volumes ranging from 50 to 200μL.

  • Comprehensive line of pipette tips for automated liquid handling systems
  • Manufactured to stringent specifications under strict process controls
  • Assembled via automation to ensure consistent performance and quality
  • Tips are certified RNase-/DNase-free and nonpyrogenic
  • Available in a range of volumes: 50, 100, 180 and 200μL
  • Compatible with the following:
    • Tecan™ Freedom Evo™, TeMo and Aquarius with MCA96 head
    • Zymark™/Caliper RapidPlate™, SciClone™ and Zephyr™ with HV96 head
    • Biotek™ Precision™ and Precision XS 8 or 12 head
    • Dynamic Devices Oasis LM 8/96-Channel-Standard Vol and 96-Channel-Low Vol head
TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 200 μL
Filtered Non-filtered
Sterility Non-sterile
Color Clear
Rack Color Light Gray
Certifications/Compliance ANSI
Clean Claims DNase, Rnase Free
For Use With (Application) Non-pyrogenic, used for stringent specifications under strict process controls
Format Rack
Tip Style Automation, Round
Packaging SBS Rack
Quantity 50 Racks/Cs.
For Use With (Equipment) Tecan Freedom EVO, Tecan TeMo, Tecan Aquarius, Zymark/Caliper RapidPlate, SciClone, Zephyr, BioTek Precision, BioTek Precision XS, Dynamic Devices Oasis LM
Material Polypropylene
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.