Learn More
Thermo Scientific™ Automation Tips for the Eppendorf™ epMotion™ Automated Pipetting System
Shop All Thermo Scientific Products

Description
Thermo Scientific™ Automation Tips for the Eppendorf™ epMotion™ Automated Pipetting System are available in non-sterile, sterile and sterile filtered tips. Each pipet tip undergoes a rigorous 15-point quality control program to ensure consistency, thus providing accurate and precise performance achieved by superior tip straightness and a low coefficient of variation. The pipette tips are validated on the actual workstations for which they are created and the sterile pipette tips are certified free of RNase, DNase, DNA, endotoxin, bioburden and pyrogen.
Features:
- Available in nonsterile, sterile and sterile filtered
- Validated on actual workstation for which tip is created
- Low percentage coefficient of variation
- Sterile tips are certified free of RNase, DNase, DNA, endotoxin, bioburden and pyrogen
Specifications
Specifications
| Volume (Metric) | 300 μL |
| Filtered | Filtered |
| Sterility | Sterile |
| Color | Clear |
| Format | Rack |
| Tip Style | Automation |
| For Use With (Equipment) | Eppendorf epMotion |
| Product Line | Eppendorf epMotion |
| Fit for Purpose | Sterile and PCR Ready |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.