Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Automation Tips for the Eppendorf™ epMotion™ Automated Pipetting System
GREENER_CHOICE

Catalog No. 14754208 Shop All Thermo Scientific Products
Change view
Click to view available options
Volume (Metric):
50 μL
300 μL
1000 μL
Filtered:
Filtered
Non-filtered
Sterility:
Non-sterile
Sterile
Fit for Purpose:
Certified Quality
Sterile
Sterile and PCR Ready
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Filtered Sterility Fit for Purpose
14-754-208 1000 μL Non-filtered Sterile Sterile
14-754-201 50 μL Filtered Sterile Sterile and PCR Ready
14-754-202 50 μL Non-filtered Sterile Sterile
14-754-203 50 μL Non-filtered Non-sterile Certified Quality
14-754-204 300 μL Filtered Sterile Sterile and PCR Ready
14-754-205 300 μL Non-filtered Sterile Sterile
14-754-206 300 μL Non-filtered Non-sterile Certified Quality
14-754-207 1000 μL Filtered Sterile Sterile and PCR Ready
14-754-209 1000 μL Non-filtered Non-sterile Certified Quality
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. 14-754-208 Supplier Thermo Scientific™ Supplier No. 967261
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Increase pipetting performance with these automation tips. Each pipette tip undergoes a rigorous 15-point quality control program to ensure consistency and accuracy achieved by superior tip straightness and a low coefficient of variation.

Thermo Scientific™ Automation Tips for the Eppendorf™ epMotion™ Automated Pipetting System are available in non-sterile, sterile and sterile filtered tips. Each pipet tip undergoes a rigorous 15-point quality control program to ensure consistency, thus providing accurate and precise performance achieved by superior tip straightness and a low coefficient of variation. The pipette tips are validated on the actual workstations for which they are created and the sterile pipette tips are certified free of RNase, DNase, DNA, endotoxin, bioburden and pyrogen.

Features:

  • Available in nonsterile, sterile and sterile filtered
  • Validated on actual workstation for which tip is created
  • Low percentage coefficient of variation
  • Sterile tips are certified free of RNase, DNase, DNA, endotoxin, bioburden and pyrogen
TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 1000 μL
Filtered Non-filtered
Sterility Sterile
Color Clear
Format Rack
Tip Style Automation
For Use With (Equipment) Eppendorf epMotion
Product Line Eppendorf epMotion
Fit for Purpose Sterile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.