Learn More
Apexbio Technology LLC Autophagy/Cytotoxicity Dual Staining Kit (MDC/PI) 2000tests
SDP
Catalog No. 5000097191
Encompass_Preferred
Autophagy Cytotoxicity Dual Staining Kit MDC PI is a dual-fluorescent dye kit designed to assess autophagic activity and cytotoxicity in live cells It employs monodansylcadaverine (MDC) which labels autophagic vacuoles by associating with acidic lipid-containing compartments and propidium iodide (PI) which stains cells with compromised membranes indicative of severe injury or cell death Autophagy Cytotoxicity Dual Staining Kit MDC PI enables simultaneous detection of autophagy and cytotoxicity by fluorescent staining In cell-based studies it allows real-time visualization and analysis of autophagic activity and cell membrane integrity Based on these properties Autophagy Cytotoxicity Dual Staining Kit MDC PI is suitable for research applications in autophagy cytotoxicity pharmacological assessment and mechanistic studies related to disease models
Catalog No. 50-000-97191 Supplier Apexbio Technology LLC Supplier No. K22882000
Autophagy Cytotoxicity Dual Staining Kit MDC PI is a dual-fluorescent dye kit designed to assess autophagic activity and cytotoxicity in live cells It employs monodansylcadaverine (MDC) which labels autophagic vacuoles by associating with acidic lipid-containing compartments and propidium iodide (PI) which stains cells with compromised membranes indicative of severe injury or cell death Autophagy Cytotoxicity Dual Staining Kit MDC PI enables simultaneous detection of autophagy and cytotoxicity by fluorescent staining In cell-based studies it allows real-time visualization and analysis of autophagic activity and cell membrane integrity Based on these properties Autophagy Cytotoxicity Dual Staining Kit MDC PI is suitable for research applications in autophagy cytotoxicity pharmacological assessment and mechanistic studies related to disease models