Learn More
Selleck Chemical LLC Autophagy Compound Library 100uL/well
SDP
Supplier:  Selleck Chemical LLC L2600100UL/WELL
Encompass_Preferred
A unique collection of 1029 small molecules with autophagy-inducing or autophagy-inhibiting activity. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8190
Only null left
Add to Cart
Request a Quote