Learn More
Enzo Life Sciences SCREEN-WELL Autophagy library (1 Library)
SDP
Catalog No. NC1624808
Encompass_Preferred
Structurally and mechanistically different compounds for studying the roles of pro- and anti-autophagic molecules in cells and .in vitro applications. Contents: 94 compounds with defined autophagy-inducing or -inhibitory activity.  A variety of structurally and mechanistically different compound classes are included. Application: HTS. Use/Stability: Stable for at least one year from the date of receipt when stored at -80°C. Long Term Storage: -80°C
Catalog No. NC1624808 Supplier Enzo Life Sciences Supplier No. BML28370100
Structurally and mechanistically different compounds for studying the roles of pro- and anti-autophagic molecules in cells and .in vitro applications. Contents: 94 compounds with defined autophagy-inducing or -inhibitory activity.  A variety of structurally and mechanistically different compound classes are included. Application: HTS. Use/Stability: Stable for at least one year from the date of receipt when stored at -80°C. Long Term Storage: -80°C