Learn More
Apexbio Technology LLC Autophagy Staining Kit (MDC) 2000tests
SDP
Supplier:  Apexbio Technology LLC K22872000
Encompass_Preferred
Autophagy Staining Kit MDC is a fluorescence-based assay kit designed to monitor autophagic activity in living cells It enables detection of autophagic vacuoles by utilizing monodansylcadaverine (MDC) a cell-permeable probe that selectively accumulates in autophagic compartments due to its affinity for phospholipids in acidic environments reflecting the autophagy process Autophagy Staining Kit MDC exerts its biological activity through the selective labeling of autophagic vacuoles in living cultured cells Based on these properties Autophagy Staining Kit MDC holds research potential in studies of autophagy-related mechanisms and diseases including neurodegenerative disorders metabolic dysfunction cancer and infectious diseases
Catalog No. 50-000-97129