Learn More
Cayman Chemical AutophagyFocused Screeng L
SDP
Catalog No. 502994768
Encompass_Preferred
An a2C-and a1D-adrenoceptor antagonist (pKis 6 54 and 8 2 respectively) and 5-HT1A partial agonist (pKi 8 3)
Catalog No. 50-299-4768 Supplier Cayman Chemical Supplier No. 23537100 UL
An a2C-and a1D-adrenoceptor antagonist (pKis 6 54 and 8 2 respectively) and 5-HT1A partial agonist (pKi 8 3)