Learn More
Selleck Chemical LLC Autophinib 5mg 1644443-47-9
SDP
Supplier:  Selleck Chemical LLC S85965MG
Encompass_Preferred
Autophinib is a potent autophagy inhibitor with a novel chemotype with IC50 values of 90 and 40 nM for autophagy in starvation induced autophagy assay and rapamycin induced autophagy assay. The IC50 value for Vps34 is 19 nM in vitro. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8106