Learn More
TA Instruments AutoPilot, Script Creation, Routine Operations Automation, Conditional Decision Making, Video and Dialog Box Guidance, OneTouch Interface
Supplier:  TA Instruments 402046.901
Encompass_Preferred
Waters TA Instruments AutoPilot adds functionality to TRIOS software, allowing users to create scripts that replicate tasks for efficiency. A script automates tasks typically performed one-by-one by an operator. AutoPilot guides new operators with videos, timers, and interactive dialog boxes, automates operations such as data analysis and reporting, and integrates conditional decision making into procedures. Scripts can be deployed across multiple laboratories to standardize sample loading, data analysis, and reporting. The OneTouch interface allows novice operators to run complex scripts with a single click, increasing efficiency and reducing errors.
Catalog No. 50-005-80817