Learn More
Electron Microscopy Sciences EMS Autopsy Grossing Knife, 14" blade
SDP
Catalog No. 501931606
Encompass_Preferred
Our Sani-Safe autopsy grossing knives have plastic non-slip safety handles that comfortably mold to your hand to reduce fatigue, as well as slip guards to protect your hands from the cutting edge. The handle is securely bonded to the cutting blade. The blade is well-balanced, hand-ground and honed, making it the perfect autopsy knife. It can be constantly sharpened for long life and easily sanitized with any disinfectant. These knives are made with premium grade stainless steel. Blade is honed with air-gap grooves.
Blade: 14"
Catalog No. 50-193-1606 Supplier Electron Microscopy Sciences Supplier No. 7206004
Our Sani-Safe autopsy grossing knives have plastic non-slip safety handles that comfortably mold to your hand to reduce fatigue, as well as slip guards to protect your hands from the cutting edge. The handle is securely bonded to the cutting blade. The blade is well-balanced, hand-ground and honed, making it the perfect autopsy knife. It can be constantly sharpened for long life and easily sanitized with any disinfectant. These knives are made with premium grade stainless steel. Blade is honed with air-gap grooves.
Blade: 14"