Learn More
World Precision Instrument Autopsy Saw, Round Tip, Bone Cutting, 30.5cm
SDP
Catalog No. 502538369
Encompass
The autopsy saw is for cutting bones in post-mortem exams. The end it rounded to reduce trauma to surrounding tissue. This saw is crafted from standard grade stainless steel, suitable for all forms of disinfection and autoclaving.
Catalog No. 50-253-8369 Supplier World Precision Instrument Supplier No. WP0149
The autopsy saw is for cutting bones in post-mortem exams. The end it rounded to reduce trauma to surrounding tissue. This saw is crafted from standard grade stainless steel, suitable for all forms of disinfection and autoclaving.