Learn More
Santa Cruz Biotechnology UltraCruz Autoradiography Film | 100/case
Catalog No. NC1734511
Encompass
Autoradiography (X-ray) film is tinted with the classic blue color, suitable for all common uses including radioisotopes and chemiluminescent substrates.
Catalog No. NC1734511 Supplier Santa Cruz Biotechnology Supplier No. SC201696
Autoradiography (X-ray) film is tinted with the classic blue color, suitable for all common uses including radioisotopes and chemiluminescent substrates.