Learn More
Research Products International Corp Autoradiography Ruler Set
SDP
Supplier:  Research Products International Corp ARS450
Encompass_Preferred
Ideal for orientation and measurement of electrophoretic markers. Exposes sharp metric scale on x-ray film for accurate and fast alignment of gel, blot and film. Also simplifies and improves accuracy of mobility calculations. High resolution phosphorescent image remains unaffected by intensifying screens. Ruler can be reused indefinitely.
Catalog No. 50-212-927