Learn More
Electron Microscopy Sciences Autoradiography Ruler Set, 20 cm length, 4/pk
SDP
Catalog No. 5034181
Encompass_Preferred
Phosphorescent, Non-Radioactive Metric Rulers for Radiographs The Autoradiography Ruler Set (ARS) is an orientation and measurement device that exposes metric ruler markings directly onto X-ray film Ideal- Utilizes a specially formulated phosphorescent dye compound and a unique emission filter which serves as an ideal non-radioactive substitute for ink markings
Easy- Make easy electrophoretic mobility calculations
Re-usable- The phosphorescent emission of each ruler can be regenerated indefinitely and can be reused any number of times
High Resolution - Produces a high resolution image that is unaffected by intensifying screens or pre-flashed film 20cm length rulers.
Catalog No. 50-341-81 Supplier Electron Microscopy Sciences Supplier No. 6855004
Phosphorescent, Non-Radioactive Metric Rulers for Radiographs The Autoradiography Ruler Set (ARS) is an orientation and measurement device that exposes metric ruler markings directly onto X-ray film Ideal- Utilizes a specially formulated phosphorescent dye compound and a unique emission filter which serves as an ideal non-radioactive substitute for ink markings
Easy- Make easy electrophoretic mobility calculations
Re-usable- The phosphorescent emission of each ruler can be regenerated indefinitely and can be reused any number of times
High Resolution - Produces a high resolution image that is unaffected by intensifying screens or pre-flashed film 20cm length rulers.