Learn More
Perkin Elmer US LLC Autosampler Aluminum Sample Pans - 10 µL, Qty. 400
Catalog No. 502112425
Encompass_Preferred
These volatile pans and covers are suitable for use with DSC autosamplers, but are also suitable for use with non-robotic systems. Most of these pans and covers can be crimped using the Universal Crimper (B0139005).
Catalog No. 50-211-2425 Supplier Perkin Elmer US LLC Supplier No. B0143015
These volatile pans and covers are suitable for use with DSC autosamplers, but are also suitable for use with non-robotic systems. Most of these pans and covers can be crimped using the Universal Crimper (B0139005).