Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Supelco™ Autosampler crimp top vials

Catalog No. 111011708 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Color:
Amber
Clear
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
11-101-1708 Amber
11-101-1868 Clear
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-101-1708 Supplier MilliporeSigma™ Supelco™ Supplier No. Z2915871PAK
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Crimp top vials constructed of borosilicate glass

  • Constructed of borosilicate glass
  • Vials use 11 mm aluminum caps
  • Order separately
TRUSTED_SUSTAINABILITY

Specifications

Color Amber
Material Glass
No. per Pack 100
Size (Metric) Opening 12 x 32 mm standard opening
Volume (English) 2 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.