Learn More
Perkin Elmer US LLC Autosampler Platinum Pan Kit for TGA 8000 - Qty. 5
Supplier:  Perkin Elmer US LLC N5370195
Encompass_Preferred
PerkinElmer's sample pans and covers are made from high quality aluminum, ceramic, or platinum. They are designed to hold a variety of substances, both volatile and non-volatile, and to withstand temperature ranging from sub-ambient to over 1000°C.
Catalog No. 50-211-1852
Add to Cart
Request a Quote