Learn More
SCP Science Autosampler Tube Caps | Clear | 1000/pk
Catalog No. NC1845350
Encompass
Clear autosampler tube caps designed to fit 16 x 100 mm and 17 x 100 mm tubes.They are manufactured from low-density polyethylene (LDPE) with ultra-low metal content for use in ICP/ICP-MS analysis.
Catalog No. NC1845350 Supplier SCP Science Supplier No. 010515672
Clear autosampler tube caps designed to fit 16 x 100 mm and 17 x 100 mm tubes.They are manufactured from low-density polyethylene (LDPE) with ultra-low metal content for use in ICP/ICP-MS analysis.