Learn More
Agilent BioTek AutoScratch Wound Making Tool
GSA_VAGREENER_CHOICE
Catalog No. 11120685 Shop All Agilent Technologies Products
Change view
Click to view available options
For Use With (Equipment):
Lionheart FX, Cytation 5, Cytation 1
Catalog No. For Use With (Equipment)
11-120-685 Lionheart FX, Cytation 5, Cytation 1
1 options
Catalog No. 11-120-685 Supplier Agilent BioTek Supplier No. AUTOSCRATCHS
Add to Cart
Add to Cart
Request a Quote
Automatically creates reproducible scratch wounds in cell monolayers grown in 24- or 96-well microplates for cell migration and invasion studies.
  • Automatically creates reproducible scratch wounds in cell monolayers grown in microplates.
  • The simple push button operation and tool-free scratch pin manifold exchange make it easy to process either 96 or 24 well plates which are commonly used in migration and invasion assays.
  • The compact system features an onboard pre-programmed cleaning routine to keep the scratch pins free of buildup and avoiding contamination.
  • AutoScratch precisely and efficiently automates the sample prep for imaging workflows with Agilent BioTeks Cytation Cell Imaging Multi-Mode Readers and Lionheart Automated Microscopes.
  • For complete capture through data publication workflows pre-defined protocols in Gen5 software automatically calculate wound width wound % confluence and maximum wound healing rate.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) Wound Healing and Cell Migration Assays
For Use With (Equipment) Lionheart FX, Cytation 5, Cytation 1