Learn More
Gilson Company Inc GilSonic AutoSiever Replacement Fines Collector
Catalog No. 50871366
Encompass_Preferred
GilSonic AutoSiever Replacement Fines Collector is a component of the GAA-2 GilSonic Autosiever Stack Assembly and required for use with the GA-6 GilSonic AutoSiever Sonic Sifter and other Sonic Sifters. The Fines Collector is made of latex. It seals the bottom of the sieve stack and collects materials passing the finest sieve. The collector's latex walls flex as a result of audio energy from the Autosiever and create a pulsing column of air. One unit is included with the AutoSiever; it is also available as a replacement item. Features: Seals sieve stack, Collects fines during testing. Included Items: Gilsonic AutoSiever Replacement Fines Collector.
Catalog No. 50-871-366 Supplier Gilson Company Inc Supplier No. GAA4
GilSonic AutoSiever Replacement Fines Collector is a component of the GAA-2 GilSonic Autosiever Stack Assembly and required for use with the GA-6 GilSonic AutoSiever Sonic Sifter and other Sonic Sifters. The Fines Collector is made of latex. It seals the bottom of the sieve stack and collects materials passing the finest sieve. The collector's latex walls flex as a result of audio energy from the Autosiever and create a pulsing column of air. One unit is included with the AutoSiever; it is also available as a replacement item. Features: Seals sieve stack, Collects fines during testing. Included Items: Gilsonic AutoSiever Replacement Fines Collector.