Learn More
Gilson Company Inc AUTOSIVER FOR FINE POWDERS
Catalog No. 50871363
Encompass_Preferred
Sieve Shaker; GilSonic AutoSiever; Dual tapper; 3000 pulses/min.; For dry sieving of fine powders from #20 to 5um; Programmable time, vibration and tapping sequences; Memory for 10 programs; Uses 3 in. acrylic sieves
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
Catalog No. 50-871-363 Supplier Gilson Company Inc Supplier No. GA6
Add to Cart
Add to Cart
Request a Quote
Sieve Shaker; GilSonic AutoSiever; Dual tapper; 3000 pulses/min.; For dry sieving of fine powders from #20 to 5um; Programmable time, vibration and tapping sequences; Memory for 10 programs; Uses 3 in. acrylic sieves
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.