Learn More
IMEB, Inc Tek-Select Autoslip Coverslips 10 per Case, 24x50, Autoslips Coverslips Specifically Designed for Instruments That Automatically Apply Cover Glass, Slides Have a No. 1 Mm Thickness
SDP
Catalog No. 501689498
Encompass_Preferred
Tek-Select Autoslip Coverslips 10 Per Case, 24x50. Our Autoslips coverslips are specifically designed for use on instruments that automatically apply cover glass, but they also perform well for manual coverslipping in humid environments. These easy-to-use coverslips contain a desiccant and are made to reduce sticking and jamming in coverslippers. Slides have a No. 1 mm thickness.
Catalog No. 50-168-9498 Supplier IMEB, Inc Supplier No. AS2450
Tek-Select Autoslip Coverslips 10 Per Case, 24x50. Our Autoslips coverslips are specifically designed for use on instruments that automatically apply cover glass, but they also perform well for manual coverslipping in humid environments. These easy-to-use coverslips contain a desiccant and are made to reduce sticking and jamming in coverslippers. Slides have a No. 1 mm thickness.