Learn More
Waters Corp Waters Standards Test Kit, Mass Spectrometry, System Assessment, 11 Components
Catalog No. 50201682 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Mass spectrometry standards kit for system assessment. It contains 11 components and is designed for the AutoSpec Premier instrument. This kit ensures reliable system performance with a shelf life of 6 months, and ships under ambient conditions.
  • Contains 11 components for comprehensive system assessment.
  • Includes essential consumables like vials, pipets, and a pipet bulb.
  • Provides a document for kit usage and a certificate of analysis.
  • Features gas chromatography (GC) and mass spectrometry (MS) detection.
  • Liquid formulation with a 6-month shelf life.
  • Suitable for room temperature storage and ambient shipping.
  • Designed as a reference type for system performance applications.
  • Packaged as a single unit.
  • Contains hazardous material requiring special freight handling.
Catalog No. 50-201-682 Supplier Waters Corp Supplier No. 700004021
Mass spectrometry standards kit for system assessment. It contains 11 components and is designed for the AutoSpec Premier instrument. This kit ensures reliable system performance with a shelf life of 6 months, and ships under ambient conditions.
  • Contains 11 components for comprehensive system assessment.
  • Includes essential consumables like vials, pipets, and a pipet bulb.
  • Provides a document for kit usage and a certificate of analysis.
  • Features gas chromatography (GC) and mass spectrometry (MS) detection.
  • Liquid formulation with a 6-month shelf life.
  • Suitable for room temperature storage and ambient shipping.
  • Designed as a reference type for system performance applications.
  • Packaged as a single unit.
  • Contains hazardous material requiring special freight handling.