Learn More
Electron Microscopy Sciences EMS Autospy Dissecting Knife Standard Stainless Steel 9⅞" Blade, 0.75" width
SDP
Catalog No. 501931588
Encompass_Preferred
All-purpose autopsy dissecting knife with a sleek and grooved handle to provide a comfortable and secure grip. Made of stainless steel, these knives feature ridged, tapered blades with pointed tips.
Length: 9⅞"
Width: 0.75"
Catalog No. 50-193-1588 Supplier Electron Microscopy Sciences Supplier No. 7205907
All-purpose autopsy dissecting knife with a sleek and grooved handle to provide a comfortable and secure grip. Made of stainless steel, these knives feature ridged, tapered blades with pointed tips.
Length: 9⅞"
Width: 0.75"