Learn More
Perkin Elmer US LLC AutoSystem Autosampler Syringe, Metal Plunger in Barrel Type, Volume: 50µL, Outer Diameter: 0.63
Catalog No. 502111528
Encompass_Preferred
Recommended autosampler syringes are available in 0.5, 5 and 50 µL capacities. For routine analyses, the metal plunger in barrel with PTFE-tipped seal is the standard syringe as shipped with each Clarus GC instrument. Alternative syringes to use are the metal plunger in barrel or the 0.53 mm on-column injection.
Catalog No. 50-211-1528 Supplier Perkin Elmer US LLC Supplier No. N6101760
Recommended autosampler syringes are available in 0.5, 5 and 50 µL capacities. For routine analyses, the metal plunger in barrel with PTFE-tipped seal is the standard syringe as shipped with each Clarus GC instrument. Alternative syringes to use are the metal plunger in barrel or the 0.53 mm on-column injection.