Learn More
Echelon Biosciences Research Labs High Throughput Autotaxin Inhibitor Screening Kit
SDP
Catalog No. 50143988
Encompass
The Autotaxin Inhibitor Screening Kit utilizes purified autotaxin (ATX) and the autotaxin substrate FS-3, which fluoresces upon cleavage. The assay is performed in a 96 well plate and quantifies the autotaxin activity in the presence of the users samples by measuring changes in fluorescence over time.Sample Type: Inhibitors against ATXSample Volume: 10 L/ well of your sample compoundsSample Considerations: Some organic solvents interfere with the assay (see technical data sheet). This item is non returnable
This item is not returnable. View return policy
Catalog No. 50-143-988 Supplier Echelon Biosciences Research Labs Supplier No. K4200HTSEAKIT
The Autotaxin Inhibitor Screening Kit utilizes purified autotaxin (ATX) and the autotaxin substrate FS-3, which fluoresces upon cleavage. The assay is performed in a 96 well plate and quantifies the autotaxin activity in the presence of the users samples by measuring changes in fluorescence over time.Sample Type: Inhibitors against ATXSample Volume: 10 L/ well of your sample compoundsSample Considerations: Some organic solvents interfere with the assay (see technical data sheet). This item is non returnable