Learn More
Medchemexpress LLC HY-135053 5mg Medchemexpress, Autotaxin-IN-3 CAS:2156655-68-2 Purity:>98%
SDP
Catalog No. 501969858
Encompass_Preferred
Medchemexpress, HY-135053 5mg Autotaxin-IN-3 CAS:2156655-68-2 Autotaxin-IN-3 is a Autotaxin(ATX) inhibitor with an IC50 of 2.4 nM, compound 33, sourced from patent WO2018212534A1. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9858 Supplier Medchemexpress LLC Supplier No. HY1350535MG
Add to Cart
Add to Cart
Medchemexpress, HY-135053 5mg Autotaxin-IN-3 CAS:2156655-68-2 Autotaxin-IN-3 is a Autotaxin(ATX) inhibitor with an IC50 of 2.4 nM, compound 33, sourced from patent WO2018212534A1. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.