Learn More
Echelon Biosciences Research Labs Autotaxin Inhibitor Screening Kit
SDP
Catalog No. 50720453
Encompass
The Autotaxin Inhibitor Screening Kit utilizes purified autotaxin (ATX) and the autotaxin substrate FS-3, which fluoresces upon cleavage. The assay is performed in a 96 well plate and quantifies the autotaxin activity in the presence of the users samples by measuring changes in fluorescence over time.Sample Type: Inhibitors against ATXSample Volume: 10 L/ well of your sample compoundsSample Considerations: Some organic solvents interfere with the assay (see technical data sheet). This item is non returnable
This item is not returnable. View return policy
Catalog No. 50-720-453 Supplier Echelon Biosciences Research Labs Supplier No. K4200EAKIT
Add to Cart
Add to Cart
The Autotaxin Inhibitor Screening Kit utilizes purified autotaxin (ATX) and the autotaxin substrate FS-3, which fluoresces upon cleavage. The assay is performed in a 96 well plate and quantifies the autotaxin activity in the presence of the users samples by measuring changes in fluorescence over time.Sample Type: Inhibitors against ATXSample Volume: 10 L/ well of your sample compoundsSample Considerations: Some organic solvents interfere with the assay (see technical data sheet). This item is non returnable