Learn More
Medchemexpress LLC Autotaxin modulator 1 | 1548743-69-6 | 99.4% | 10 MG
SDP
Catalog No. 5000340925
Encompass_Preferred
Autotaxin modulator 1 is an autotaxin (ATX) enzyme inhibitor, extracted from patent WO 2014018881 A1, Compound Example 12b. It is expected to be useful for researching demyelination due to injury or disease, as well as for researching proliferative disorders such as cancer.
  • Autotaxin (ATX) enzyme inhibitor
  • Useful for researching demyelination due to injury or disease
  • Useful for researching proliferative disorders such as cancer
  • For research use only
  • Appears as a white to off-white solid
  • Soluble in DMSO (100 mg/mL)
Catalog No. 50-003-40925 Supplier Medchemexpress LLC Supplier No. HY1281210MG
Autotaxin modulator 1 is an autotaxin (ATX) enzyme inhibitor, extracted from patent WO 2014018881 A1, Compound Example 12b. It is expected to be useful for researching demyelination due to injury or disease, as well as for researching proliferative disorders such as cancer.
  • Autotaxin (ATX) enzyme inhibitor
  • Useful for researching demyelination due to injury or disease
  • Useful for researching proliferative disorders such as cancer
  • For research use only
  • Appears as a white to off-white solid
  • Soluble in DMSO (100 mg/mL)