Learn More
Medchemexpress LLC HY-12812 5mg Medchemexpress, Autotaxin modulator 1 CAS:1548743-69-6 Purity:>98%
SDP
Catalog No. 502081151
Encompass_Preferred
Medchemexpress, HY-12812 5mg Autotaxin modulator 1 CAS:1548743-69-6 Autotaxin modulator 1 is an autotaxin (ATX) enzyme inhibitor, extracted from patent WO 2014018881 A1, Compound Example 12b. Autotaxin modulator 1 is expected to be useful for researching demyelination due to injury or disease, as well as for researching proliferative disorders such as cancer. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1151 Supplier Medchemexpress LLC Supplier No. HY128125MG
Add to Cart
Add to Cart
Medchemexpress, HY-12812 5mg Autotaxin modulator 1 CAS:1548743-69-6 Autotaxin modulator 1 is an autotaxin (ATX) enzyme inhibitor, extracted from patent WO 2014018881 A1, Compound Example 12b. Autotaxin modulator 1 is expected to be useful for researching demyelination due to injury or disease, as well as for researching proliferative disorders such as cancer. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.