Learn More
Echelon Biosciences Research Labs FS-3 (LysoPLD / Autotaxin substrate) 200 ug
SDP
Catalog No. 50720497
Encompass
Echelons fluorogenic Autotaxin (ATX) substrate FS-3 (L-2000) is a doubly labeled analog of LPC wherin the fluorophore is quenched through intramolecular energy transfer. Hydrolysis of the substrate produces an increase in fluorescence. This item is non returnable
This item is not returnable. View return policy
Catalog No. 50-720-497 Supplier Echelon Biosciences Research Labs Supplier No. L2000200UG
Echelons fluorogenic Autotaxin (ATX) substrate FS-3 (L-2000) is a doubly labeled analog of LPC wherin the fluorophore is quenched through intramolecular energy transfer. Hydrolysis of the substrate produces an increase in fluorescence. This item is non returnable