Learn More
Hach Company Aux. Cable Assembly Adaptor
Supplier:  Hach Company 8758000
Encompass
Auxiliary Cable Assembly Adaptor with Cap6 to 7 pin auxiliary cable adaptor allows use of 6 pin auxiliary cable and 7 pin SD900 sampler controller
Catalog No. 50-002-07093