Learn More
Waters Corp Auxillary Valve Cartridge Kit, Convergence Manager, Injector/Valve Pod, UPC2 System
Catalog No. 502390995 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This replacement valve cartridge kit is designed for the Convergence Manager. The kit provides inlet and outlet tubing assemblies and is specifically used in UPC2 systems.
  • Provides inlet and outlet tubing assemblies
  • Used in UPC2 systems
  • Type: Injector/Valve Pod
  • Units per package: 1
  • Recommended for annual replacement
Catalog No. 50-239-0995 Supplier Waters Corp Supplier No. 700009052
This replacement valve cartridge kit is designed for the Convergence Manager. The kit provides inlet and outlet tubing assemblies and is specifically used in UPC2 systems.
  • Provides inlet and outlet tubing assemblies
  • Used in UPC2 systems
  • Type: Injector/Valve Pod
  • Units per package: 1
  • Recommended for annual replacement